Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-20
Entrez Gene ID
189730
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_Y39B6A.46
Chromosome Location
Chr.V: 18957578 - 18960172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171561.4 NP_741651.1 2 - 3 195 125

View Full Product Details