Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-46
Entrez Gene ID
180252
Gene Name
protein-coding
Gene Aliases
CELE_Y39B6A.19
Chromosome Location
Chr.V: 19060173 - 19067034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136444.3 NP_001129916.1 3 - 4 569 118
NM_171588.6 NP_741678.1 4 - 5 579 118

View Full Product Details