Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifc-2
Entrez Gene ID
180414
Gene Name
protein-coding
Gene Aliases
CELE_M6.1
Chromosome Location
Chr.X: 642156 - 654939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374936.2 NP_001362085.1 13 - 14 2192 81
NM_001374938.1 NP_001362002.1 14 - 15 2265 81
NM_001380893.1 NP_001367765.1 10 - 11 1973 81

View Full Product Details