Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-40
Entrez Gene ID
180560
Gene Name
protein-coding
Gene Aliases
CELE_T03G6.2
Chromosome Location
Chr.X: 2594003 - 2606732 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029702.5 NP_001024873.1 10 - 11 1150 116
NM_001374979.2 NP_001362027.1 10 - 11 1125 116

View Full Product Details