Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flp-11
Entrez Gene ID
180741
Gene Name
protein-coding
Gene Aliases
CELE_K02G10.4
Chromosome Location
Chr.X: 4677334 - 4678334 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029581.9 NP_001024752.1 1 - 2 133 58
NM_001029582.3 NP_001024753.1 2 - 3 278 58
NM_001029583.4 NP_001024754.1 1 - 2 77 58
GenBank mRNA AF042397.1 - 1 - 2 77 58
AF042398.1 - 1 - 2 77 58

View Full Product Details