Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ags-3
Entrez Gene ID
180801
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.4
Chromosome Location
Chr.X: 5289728 - 5303600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322618.4 NP_001309689.1 7 - 8 841 87
NM_001322619.4 NP_001309690.1 6 - 7 712 87
NM_001322621.4 NP_001309692.1 1 - 2 76 87
NM_001383577.1 13 - 14 1727 87
NM_171684.6 NP_741787.1 13 - 14 1651 87
NR_136376.1 13 - 14 1711 87
NR_136377.1 11 - 12 1557 87

View Full Product Details