Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bca-1
Entrez Gene ID
180890
Gene Name
Beta carbonic anhydrase 1
Gene Aliases
CELE_T13C5.5
Chromosome Location
Chr.X: 6201874 - 6206408 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171701.6 NP_741809.1 4 - 5 1076 81

View Full Product Details