Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-1
Entrez Gene ID
180968
Gene Name
protein-coding
Gene Aliases
CELE_C16E9.4
Chromosome Location
Chr.X: 6943735 - 6956399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029236.3 NP_001024407.1 9 - 10 1228 84
NM_171716.5 NP_741826.1 9 - 10 1228 84
NR_131721.1 9 - 10 1172 84
NR_131722.1 9 - 10 1172 84
NR_131723.1 8 - 9 1152 84
NR_131724.1 8 - 9 1152 84
GenBank mRNA U59212.1 - 9 - 10 1228 84

View Full Product Details