Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flp-26
Entrez Gene ID
259712
Gene Name
GGAGEPLAFSPDMLSLRF-amide
Gene Aliases
CELE_R173.4
Chromosome Location
Chr.X: 7029136 - 7029638 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171717.6 NP_741827.1 1 - 2 111 88

View Full Product Details