The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_171727.4 | NP_741838.1 | 1 - 2 | 423 | 117 |
GenBank mRNA | AF082011.1 | - | 1 - 2 | 423 | 117 |