Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grd-1
Entrez Gene ID
181300
Gene Name
GRounDhog (hedgehog-like family)
Gene Aliases
CELE_R08B4.1
Chromosome Location
Chr.X: 11109440 - 11115887 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029672.3 NP_001024843.1 12 - 13 3210 77

View Full Product Details