Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eor-2
Entrez Gene ID
181557
Gene Name
protein-coding
Gene Aliases
CELE_C44H4.7
Chromosome Location
Chr.X: 14603017 - 14608660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029299.6 NP_001024470.1 8 - 9 1264 96
NM_001029300.6 NP_001024471.1 7 - 8 1219 96
NM_001313378.4 NP_001300307.1 7 - 7 1189 96
GenBank mRNA AF502570.1 - 8 - 9 1211 96
AF519109.1 - 8 - 9 1189 96
AF519110.1 - 7 - 8 1144 96

View Full Product Details