Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vap-1
Entrez Gene ID
181768
Gene Name
Venom-Allergen-like Protein
Gene Aliases
CELE_F11C7.3
Chromosome Location
Chr.X: 17393939 - 17396556 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029382.3 NP_001024553.1 4 - 5 615 93

View Full Product Details