Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-96
Entrez Gene ID
180413
Gene Name
protein-coding
Gene Aliases
CELE_F13C5.6
Chromosome Location
Chr.X: 611103 - 621290 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171939.5 NP_741704.1 10 - 11 1151 71

View Full Product Details