Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ags-3
Entrez Gene ID
180801
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.4
Chromosome Location
Chr.X: 5289728 - 5303600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383577.1 3 - 4 375 283
NR_136376.1 3 - 4 359 283

View Full Product Details