Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-80
Entrez Gene ID
175167
Gene Name
protein-coding
Gene Aliases
CELE_H10E21.3
Chromosome Location
Chr.III: 11985 - 14869 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064725.7 NP_497126.1 2 - 3 297 82
NM_181901.2 NP_871630.1 1 - 2 235 82
GenBank mRNA AY204179.1 - 2 - 3 225 82

View Full Product Details