Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-3
Entrez Gene ID
182356
Gene Name
protein-coding
Gene Aliases
CELE_C07A9.7
Chromosome Location
Chr.III: 9691027 - 9694574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066742.5 NP_499143.1 5 - 6 726 92
NM_181940.6 NP_871669.1 5 - 6 728 92

View Full Product Details