Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-19
Entrez Gene ID
191090
Gene Name
protein-coding
Gene Aliases
CELE_ZC155.5
Chromosome Location
Chr.III: 5215949 - 5217611 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181964.4 NP_871693.1 2 - 3 240 90

View Full Product Details