Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-95
Entrez Gene ID
176602
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_Y41C4A.16
Chromosome Location
Chr.III: 11703885 - 11705091 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383002.1 1 - 2 209 87

View Full Product Details