Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mei-1
Entrez Gene ID
172612
Gene Name
protein-coding
Gene Aliases
CELE_T01G9.5
Chromosome Location
Chr.I: 8294678 - 8296915 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383460.1 4 - 5 1400 80

View Full Product Details