Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ugt-31
Entrez Gene ID
189771
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_Y39G10AR.6
Chromosome Location
Chr.I: 2333298 - 2338693 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026585.5 NP_001021756.1 5 - 6 657 77

View Full Product Details