Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bpl-1
Entrez Gene ID
174039
Gene Name
protein-coding
Gene Aliases
CELE_F13H8.10
Chromosome Location
Chr.II: 6274115 - 6279720 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383861.1 12 - 13 1993 76
NM_062856.7 NP_495257.3 12 - 13 2033 76
NM_182120.5 NP_871920.1 1 - 2 171 76
GenBank mRNA AY601656.1 - 1 - 2 150 76
AY601657.1 - 1 - 2 79 76

View Full Product Details