Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-2
Entrez Gene ID
174061
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_T25D10.5
Chromosome Location
Chr.II: 6401949 - 6403105 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182195.5 NP_871995.1 1 - 2 145 138

View Full Product Details