Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-18
Entrez Gene ID
353391
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F39E9.12
Chromosome Location
Chr.II: 3298214 - 3299590 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182266.3 NP_872066.2 2 - 3 240 87

View Full Product Details