Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wnk-1
Entrez Gene ID
177743
Gene Name
protein-coding
Gene Aliases
CELE_C46C2.1
Chromosome Location
Chr.IV: 9200102 - 9212242 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268439.4 NP_001255368.1 13 - 14 5314 75
NM_001268440.3 NP_001255369.1 11 - 12 4510 75
NM_001268441.3 NP_001255370.1 13 - 14 5313 75
NM_001268442.4 NP_001255371.1 11 - 12 4501 75
NM_001268443.3 NP_001255372.1 12 - 13 4987 75
NM_001268444.3 NP_001255373.1 11 - 12 4504 75
NM_001268445.3 NP_001255374.1 10 - 11 4834 75
NM_001268446.3 NP_001255375.1 10 - 11 4825 75
NM_001268447.3 NP_001255376.1 10 - 11 4828 75
NM_001268448.3 NP_001255377.1 10 - 11 4819 75
NM_001307103.3 NP_001294032.1 13 - 14 5318 75
NM_001307104.3 NP_001294033.1 11 - 12 4510 75
NM_001307106.4 NP_001294035.1 11 - 12 4501 75
NM_001307107.3 NP_001294036.1 13 - 14 5308 75
NM_001307108.3 NP_001294037.1 11 - 12 4504 75
NM_001307109.4 NP_001294038.1 12 - 13 4812 75
NM_001307110.3 NP_001294039.1 1 - 2 43 75
NM_001307111.4 NP_001294040.1 1 - 2 43 75
NM_001380399.1 NP_001368454.1 13 - 14 5305 75
NM_001380400.1 NP_001368092.1 13 - 14 5299 75
NM_069202.6 NP_501603.3 13 - 14 5293 75
NM_182275.7 NP_872075.2 12 - 13 4812 75

View Full Product Details