Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-26
Entrez Gene ID
353424
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y73B6BL.40
Chromosome Location
Chr.IV: 6299273 - 6301864 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182313.1 NP_872113.1 4 - 5 531 152

View Full Product Details