Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-43
Entrez Gene ID
3565024
Gene Name
protein-coding
Gene Aliases
CELE_F32H5.7
Chromosome Location
Chr.V: 13346278 - 13356168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276947.3 NP_001263876.1 9 - 10 1135 146
NM_001276948.3 NP_001263877.1 9 - 10 1108 146
NM_001276949.3 NP_001263878.1 6 - 7 877 146
NM_182337.7 NP_872137.2 10 - 11 1215 146

View Full Product Details