Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-11
Entrez Gene ID
179088
Gene Name
protein-coding
Gene Aliases
CELE_F20A1.7
Chromosome Location
Chr.V: 7028258 - 7036575 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356689.2 NP_001343632.1 10 - 11 1880 98
NM_001383329.1 10 - 11 1752 98
NM_072382.9 NP_504783.2 10 - 11 1885 98

View Full Product Details