The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_060438.7 | NP_492839.4 | 7 - 8 | 862 | 91 |
GenBank mRNA | AB297384.1 | - | 7 - 8 | 848 | 91 |