Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-62
Entrez Gene ID
188991
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_T27C10.2
Chromosome Location
Chr.I: 10899089 - 10902220 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060440.5 NP_492841.3 2 - 3 293 114

View Full Product Details