Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ztf-6
- Entrez Gene ID
-
173014
- Gene Name
- Zinc finger putative Transcription Factor family
- Gene Aliases
-
CELE_W06H12.1
- Chromosome Location
-
Chr.I: 11206910 - 11211961 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060476.6 |
NP_492877.1 |
2 - 3 |
299 |
84 |
View Full Product Details