Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-6
Entrez Gene ID
173014
Gene Name
Zinc finger putative Transcription Factor family
Gene Aliases
CELE_W06H12.1
Chromosome Location
Chr.I: 11206910 - 11211961 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060476.6 NP_492877.1 4 - 5 1245 81

View Full Product Details