Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-74
Entrez Gene ID
191723
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C27C7.3
Chromosome Location
Chr.I: 11428980 - 11432156 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060518.3 NP_492919.2 5 - 6 493 87

View Full Product Details