Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spe-9
Entrez Gene ID
173042
Gene Name
protein-coding
Gene Aliases
CELE_C17D12.6
Chromosome Location
Chr.I: 11582743 - 11586069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060554.5 NP_492955.1 3 - 4 240 116
GenBank mRNA AF049327.1 - 3 - 4 218 116

View Full Product Details