Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhhc-2
Entrez Gene ID
173062
Gene Name
Palmitoyltransferase
Gene Aliases
CELE_Y47H9C.2
Chromosome Location
Chr.I: 11836460 - 11849258 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060606.6 NP_493007.2 3 - 4 490 92

View Full Product Details