Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-81
Entrez Gene ID
191728
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C47F8.8
Chromosome Location
Chr.I: 12327982 - 12329508 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060717.4 NP_493118.2 1 - 2 184 109

View Full Product Details