Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndx-1
Entrez Gene ID
173138
Gene Name
Putative nudix hydrolase 1
Gene Aliases
CELE_T26E3.2
Chromosome Location
Chr.I: 12679207 - 12681818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060808.3 NP_493209.1 5 - 6 568 63

View Full Product Details