Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sptf-3
Entrez Gene ID
189783
Gene Name
SP (Specificity Protein) Transcription Factor
Gene Aliases
CELE_Y40B1A.4
Chromosome Location
Chr.I: 13360053 - 13369972 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060952.5 NP_493353.3 7 - 8 1441 101

View Full Product Details