Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
agef-1
Entrez Gene ID
173231
Gene Name
protein-coding
Gene Aliases
CELE_Y6B3A.1
Chromosome Location
Chr.I: 13603843 - 13626225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026627.7 NP_001021798.2 17 - 18 4499 66

View Full Product Details