Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agef-1
Entrez Gene ID
173231
Gene Name
protein-coding
Gene Aliases
CELE_Y6B3A.1
Chromosome Location
Chr.I: 13603843 - 13626225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026627.7 NP_001021798.2 2 - 3 488 107
NM_001026628.5 NP_001021799.1 2 - 3 461 107

View Full Product Details