Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dkf-1
Entrez Gene ID
173234
Gene Name
Serine/threonine-protein kinase dkf-1
Gene Aliases
CELE_W09C5.5
Chromosome Location
Chr.I: 13640297 - 13648405 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060989.6 NP_493390.1 10 - 11 2123 85

View Full Product Details