Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-31
Entrez Gene ID
173235
Gene Name
protein-coding
Gene Aliases
CELE_W09C5.6
Chromosome Location
Chr.I: 13648978 - 13653704 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060990.7 NP_493391.1 1 - 2 114 74

View Full Product Details