Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsd-1
Entrez Gene ID
189371
Gene Name
3Beta_HSD domain-containing protein
Gene Aliases
CELE_Y6B3B.11
Chromosome Location
Chr.I: 13720125 - 13724453 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061001.1 NP_493402.1 4 - 5 645 65

View Full Product Details