Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taf-1
Entrez Gene ID
173257
Gene Name
Transcription initiation factor TFIID subunit 1
Gene Aliases
CELE_W04A8.7
Chromosome Location
Chr.I: 13861468 - 13887640 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061025.5 NP_493426.2 1 - 2 135 132

View Full Product Details