Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-29
Entrez Gene ID
173278
Gene Name
protein-coding
Gene Aliases
CELE_Y63D3A.6
Chromosome Location
Chr.I: 14104425 - 14113108 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383758.1 1 - 2 142 73
NM_061063.5 NP_493464.1 1 - 2 142 73

View Full Product Details