Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exoc-8
Entrez Gene ID
173320
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.2
Chromosome Location
Chr.I: 14634449 - 14642574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026531.5 NP_001021702.1 5 - 6 1067 88

View Full Product Details