Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmp-2
Entrez Gene ID
173338
Gene Name
protein-coding
Gene Aliases
CELE_K05C4.6
Chromosome Location
Chr.I: 14738882 - 14742671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265496.3 NP_001252425.1 5 - 6 1943 66
NM_001265497.3 NP_001252426.1 4 - 5 1884 66
GenBank mRNA AF016853.1 - 4 - 5 1909 66

View Full Product Details