Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
frm-1
Entrez Gene ID
173360
Gene Name
protein-coding
Gene Aliases
CELE_ZK270.2
Chromosome Location
Chr.I: 14875823 - 14900322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026695.4 NP_001021866.1 3 - 4 422 94
NM_061198.5 NP_493599.2 2 - 3 285 94
NM_061199.6 NP_493600.2 2 - 3 285 94
NM_061200.6 NP_493601.2 2 - 3 280 94

View Full Product Details