Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-1
Entrez Gene ID
173360
Gene Name
protein-coding
Gene Aliases
CELE_ZK270.2
Chromosome Location
Chr.I: 14875823 - 14900322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061200.6 NP_493601.2 8 - 9 12972 102

View Full Product Details