Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chs-2
Entrez Gene ID
173405
Gene Name
protein-coding
Gene Aliases
CELE_F48A11.1
Chromosome Location
Chr.II: 196985 - 211941 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313615.3 NP_001300544.1 11 - 12 3926 147
NM_001313616.3 NP_001300545.1 13 - 14 4208 147
NM_061281.6 NP_493682.2 11 - 12 4336 147
GenBank mRNA AY874872.1 - 11 - 12 4112 147

View Full Product Details